missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Methylmorpholine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7133471
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2847200MG 200 mg
M28471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2847200MG Supplier TCI America Supplier No. M2847200MG
Only null left

Chemical Identifiers

CAS 953780-78-4
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.607
MDL Number MFCD18382515
InChI Key MSOCQCWIEBVSLF-NUBCRITNSA-N
Synonym r-3-methylmorpholine hydrochloride, 3r-3-methylmorpholine hydrochloride, c5h11no.clh, ksc494m2h, r-3-methylmorpholin hydrochloride, r-3-methylmorpholinehydrochloride, r-3-methyl-morpholine hydrochloride, 3r-3-methyl-morpholine hydrochloride, 3r-3-methylmorpholine-hydrogen chloride 1/1
PubChem CID 57356922
IUPAC Name (3R)-3-methylmorpholine;hydrochloride
SMILES CC1COCCN1.Cl

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 200 mg
Formula Weight 137.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-3-Methylmorpholine Hydrochloride
TSCA No
Recommended Storage Refrigerator