missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Hydroxytetrahydrofuran 98.0+%, TCI America™

Catalog No. H13321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13321G 1 g
H13325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13321G Supplier TCI America Supplier No. H13321G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86087-24-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00067101
InChI Key XDPCNPCKDGQBAN-SCSAIBSYSA-N
Synonym r---3-hydroxytetrahydrofuran, r-tetrahydrofuran-3-ol, r-3-hydroxytetrahydrofuran, 3r-oxolan-3-ol, 3r-tetrahydrofuran-3-ol, r-+-3-hydroxytetrahydrofuran, r-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3r, pubchem7071, 2-furanyl,tetrahydro-3-hydroxy-, r-9ci
PubChem CID 641512
IUPAC Name (3R)-oxolan-3-ol
SMILES C1COCC1O

Specifications

Color Colorless
Quantity 1 g
Physical Form Clear Liquid at 20°C
Boiling Point 80°C
Formula Weight 88.11
Percent Purity ≥98.0% (GC)
Chemical Name or Material (R)-3-Hydroxytetrahydrofuran