missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Hydroxypyrrolidine hydrochloride, 97%, Thermo Scientific™

Catalog No. AAH5274306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5274306 5 g
AAH5274303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5274306 Supplier Thermo Scientific Chemicals Supplier No. H5274306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-3-Hydroxypyrrolidine hydrochloride is used as a darifenacin Intermediate.

Solubility
Soluble in DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 104706-47-0
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD00191570
InChI Key QPMSJEFZULFYTB-PGMHMLKASA-N
Synonym r-3-hydroxypyrrolidine hydrochloride, r---3-pyrrolidinol hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, r-pyrrolidin-3-ol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-pyrrolidinol hydrochloride, 3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride
PubChem CID 2759336
IUPAC Name (3R)-pyrrolidin-3-ol;hydrochloride
SMILES C1CNCC1O.Cl

Specifications

Melting Point 104°C to 107°C
Quantity 5 g
Beilstein 4450078
Solubility Information Soluble in DMSO.
Optical Rotation −7.6° (c=3.5 in Methanol)
Formula Weight 123.58
Percent Purity 97%
Chemical Name or Material (R)-3-Hydroxypyrrolidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures