Learn More
(R)-(+)-3-Hydroxy-3-phenylpropionic acid, 98+%
CAS: 2768-42-5 | C9H9O3 | 165.17 g/mol
Supplier: Thermo Scientific Chemicals L1904003
Description
It is employed in the synthesis and evaluation of a novel, reusable solid-supported open chain chiral auxiliary derived from m-hydrobenzoin for the diastereoselective reduction of α-keto esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed in the synthesis and evaluation of a novel, reusable solid-supported open chain chiral auxiliary derived from m-hydrobenzoin for the diastereoselective reduction of α-keto esters.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| (R)-(+)-3-Hydroxy-3-phenylpropionic acid | |
| 118°C to 121°C | |
| MFCD00145219 | |
| 3197772 | |
| AYOLELPCNDVZKZ-MRVPVSSYSA-M | |
| O[C@H](CC([O-])=O)C1=CC=CC=C1 | |
| 165.17 | |
| CHEBI:51059 | |
| ≥98% |
| 2768-42-5 | |
| C9H9O3 | |
| 1 g | |
| 3r-3-hydroxy-3-phenylpropanoic acid, r-3-hydroxy-3-phenylpropanoic acid, unii-s2w4lws09c component, r-3-hydroxy-3-phenylpropanoic acid, +-3-phenylhydracrylic acid, r-+-3-hydroxy-3-phenylpropionic acid, r-+-3-hydroxy-3-phenylpropanoic acid, benzenepropanoic acid,, r-+-3-hydroxy-3-phenylpropanoic acid, benzenepropanoic acid,, a-hydroxy-,, ar | |
| +22° (c=4 in Methanol) | |
| (3R)-3-hydroxy-3-phenylpropanoic acid | |
| 6950815 | |
| 166.18 |
Chemical Identifiers
| 2768-42-5 | |
| 165.17 | |
| AYOLELPCNDVZKZ-MRVPVSSYSA-M | |
| 6950815 | |
| (3R)-3-hydroxy-3-phenylpropanoic acid |
| C9H9O3 | |
| MFCD00145219 | |
| 3r-3-hydroxy-3-phenylpropanoic acid, r-3-hydroxy-3-phenylpropanoic acid, unii-s2w4lws09c component, r-3-hydroxy-3-phenylpropanoic acid, +-3-phenylhydracrylic acid, r-+-3-hydroxy-3-phenylpropionic acid, r-+-3-hydroxy-3-phenylpropanoic acid, benzenepropanoic acid,, r-+-3-hydroxy-3-phenylpropanoic acid, benzenepropanoic acid,, a-hydroxy-,, ar | |
| CHEBI:51059 | |
| O[C@H](CC([O-])=O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only