missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-(Dimethylamino)pyrrolidine, 98%

Catalog No. p-4090108
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426990010 1 g Glass bottle
AC426990050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426990010 Supplier Thermo Scientific Chemicals Supplier No. 426990010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 132958-72-6 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132958-72-6
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00191347
InChI Key AVAWMINJNRAQFS-ZCFIWIBFSA-N
Synonym r-3-dimethylaminopyrrolidine, r-+-3-dimethylamino pyrrolidine, r-n,n-dimethylpyrrolidin-3-amine, 3r-+-3-dimethylamino pyrrolidine, 3r-n,n-dimethylpyrrolidin-3-amine, 3r-3-dimethylaminopyrrolidine, r-3-dimethylamino pyrrolidine, r-dmap, 3r-n,n-dimethyl-3-pyrrolidinamine, r-dimethyl-pyrrolidin-3-yl-amine
PubChem CID 2758521
IUPAC Name (3R)-N,N-dimethylpyrrolidin-3-amine
SMILES CN(C)C1CCNC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Boiling Point 166°C
Flash Point 51.7°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material (R)-(+)-3-(Dimethylamino)pyrrolidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only