missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Butyn-2-ol 98.0+%, TCI America™

Catalog No. B29091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29091G 1 g
B29095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29091G Supplier TCI America Supplier No. B29091G

Chemical Identifiers

CAS 42969-65-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00211237
InChI Key GKPOMITUDGXOSB-SCSAIBSYSA-N
Synonym r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol
PubChem CID 638102
IUPAC Name (2R)-but-3-yn-2-ol
SMILES CC(C#C)O

Specifications

Color Yellow
Boiling Point 103°C
Quantity 1 g
UN Number 2929
Formula Weight 70.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-3-Butyn-2-ol