missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Butyn-2-ol, 99%

CAS: 42969-65-3 | C4H6O | 70.091 g/mol

Supplier:  Thermo Scientific Chemicals L18880MD

 View more versions of this product

Catalog No. AAL18880MD


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(R)-(+)-3-Butyn-2-ol
0.891
28°C (82°F)
1.426
250 mg
3587272
GKPOMITUDGXOSB-SCSAIBSYSA-N
CC(C#C)O
70.091
70.09
42969-65-3
108°C to 109°C
C4H6O
MFCD00211237
UN2929
r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol
+47° (Neat)
(2R)-but-3-yn-2-ol
638102
99%

Chemical Identifiers

42969-65-3
70.091
GKPOMITUDGXOSB-SCSAIBSYSA-N
638102
CC(C#C)O
C4H6O
MFCD00211237
r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol
(2R)-but-3-yn-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H300+H310+H330-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only