Learn More
(R)-(+)-3-Butyn-2-ol, 99%
CAS: 42969-65-3 | C4H6O | 70.091 g/mol
Supplier: Thermo Scientific Chemicals L1888006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (R)-(+)-3-Butyn-2-ol | |
| 0.891 | |
| 28°C (82°F) | |
| 1.426 | |
| 5 g | |
| 3587272 | |
| GKPOMITUDGXOSB-SCSAIBSYSA-N | |
| CC(C#C)O | |
| 70.091 | |
| 70.09 |
| 42969-65-3 | |
| 108°C to 109°C | |
| C4H6O | |
| MFCD00211237 | |
| UN2929 | |
| r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol | |
| +47° (Neat) | |
| (2R)-but-3-yn-2-ol | |
| 638102 | |
| 99% |
Chemical Identifiers
| 42969-65-3 | |
| 70.091 | |
| GKPOMITUDGXOSB-SCSAIBSYSA-N | |
| 638102 | |
| CC(C#C)O |
| C4H6O | |
| MFCD00211237 | |
| r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol | |
| (2R)-but-3-yn-2-ol |
Safety and Handling
GHS H Statement
H300-H310-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H300+H310+H330-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only