Learn More
(R)-(+)-3-Butyn-2-ol, 98%
CAS: 42969-65-3 | C4H6O | 70.09 g/mol
$259.19 - $259.19
Chemical Identifiers
| CAS | 42969-65-3 |
|---|---|
| Molecular Formula | C4H6O |
| Molecular Weight (g/mol) | 70.09 |
| MDL Number | MFCD00211237 |
| InChI Key | GKPOMITUDGXOSB-SCSAIBSYSA-N |
| Synonym | r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol |
| PubChem CID | 638102 |
| IUPAC Name | (2R)-but-3-yn-2-ol |
| SMILES | CC(C#C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC366300010
|
Thermo Scientific Chemicals
366300010 |
1 g | Glass bottle |
Each for $259.19
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 42969-65-3 | |
| 70.09 | |
| GKPOMITUDGXOSB-SCSAIBSYSA-N | |
| 638102 | |
| CC(C#C)O |
| C4H6O | |
| MFCD00211237 | |
| r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol | |
| (2R)-but-3-yn-2-ol |
Specifications
| 42969-65-3 | |
| 100.0 | |
| 0.8900g/mL | |
| 26°C | |
| Authentic | |
| Glass bottle | |
| 1.4235 to 1.4255 | |
| 1 g | |
| + 46.70 (20.00°C neat) | |
| r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol | |
| CC(C#C)O | |
| 70.09 | |
| 70.09 | |
| Liquid |
| 97.5 | |
| Colorless to Yellow | |
| 108.0°C to 111.0°C | |
| 97.5% min. (GC) | |
| 97.5% min. (GC) | |
| C4H6O | |
| MFCD00211237 | |
| 0.89 | |
| + 46.70 | |
| GKPOMITUDGXOSB-SCSAIBSYSA-N | |
| (2R)-but-3-yn-2-ol | |
| 638102 | |
| 98%,98% ee | |
| (R)-(+)-3-Butyn-2-ol, 98% ee |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Fatal if swallowed.
Fatal in contact with skin.
Causes serious eye irritation.
Flammable liquid and vapor.
Fatal if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
RUO – Research Use Only