missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Bromo-2-hydroxy-2-methylpropionic Acid 98.0+%, TCI America™

Catalog No. B48191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48191G 1 g
B48195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48191G Supplier TCI America Supplier No. B48191G

Chemical Identifiers

CAS 261904-39-6
Molecular Formula C4H7BrO3
Molecular Weight (g/mol) 183.00
MDL Number MFCD09751022
InChI Key HBJAYXGUOOININ-UHFFFAOYNA-N
PubChem CID 10582468
IUPAC Name 3-bromo-2-hydroxy-2-methylpropanoic acid
SMILES CC(O)(CBr)C(O)=O

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 183.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-3-Bromo-2-hydroxy-2-methylpropionic Acid