missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%

Catalog No. AAH6238103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6238103 1 g
AAH6238106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6238103 Supplier Thermo Scientific Chemicals Supplier No. H6238103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 198544-42-2 | C23H26N2O6 | 426.469 g/mol

(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 198544-42-2
Molecular Formula C23H26N2O6
Molecular Weight (g/mol) 426.469
MDL Number MFCD00798648
InChI Key PKAUMAVONPSDRW-LJQANCHMSA-N
Synonym fmoc-d-dap boc-oh, n2-fmoc-n3-boc-d-2,3-diaminopropionic acid, fmoc-n3-boc-d-2,3-diaminopropionic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, fmoc-n-beta-boc-d-alpha,beta-diaminopropionicacid, 2r-3-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1320, fmoc-d-dpr boc-oh, fmoc-d-dapa boc-oh, fmoc-dap boc-oh hydrate
PubChem CID 17040127
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 426.46
Percent Purity 95%
Physical Form Solid
Chemical Name or Material (R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only