Learn More
(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 111769-27-8 |
|---|---|
| Molecular Formula | C11H17NO4S |
| Molecular Weight (g/mol) | 259.32 |
| MDL Number | MFCD00040622,MFCD00040622,MFCD00066238 |
| InChI Key | BZXPLADBSZWDIH-UHFFFAOYNA-N |
| Synonym | r-3-aminotetrahydrofuran tosylate, r-+-tetrahydro-3-furylamine p-toluenesulfonate salt, r---3-aminotetrahydrofuran toluene-4-sulfonate, r-+-3-aminotetrahydrofuran toluene-4-sulfonate, r---3-aminotetrahydrofurantoluene-4-sulfonate, 3-furanamine, tetrahydro-, r-, 4-methylbenzenesulfonate, c4h9no.c7h8o3s, r-tetrahydrofuran-3-amine-4-methylbenzenesulfonate, 3r-oxolan-3-amine; para-toluene sulfonate, 4-methylbenzenesulfonic acid; 3r-tetrahydrofuran-3-amine |
| PubChem CID | 14243169 |
| IUPAC Name | 4-methylbenzenesulfonic acid;(3R)-oxolan-3-amine |
| SMILES | NC1CCOC1.CC1=CC=C(C=C1)S(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC435820050
|
Thermo Scientific Chemicals
435820050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 111769-27-8 | |
| 259.32 | |
| BZXPLADBSZWDIH-UHFFFAOYNA-N | |
| 14243169 | |
| NC1CCOC1.CC1=CC=C(C=C1)S(O)(=O)=O |
| C11H17NO4S | |
| MFCD00040622,MFCD00040622,MFCD00066238 | |
| r-3-aminotetrahydrofuran tosylate, r-+-tetrahydro-3-furylamine p-toluenesulfonate salt, r---3-aminotetrahydrofuran toluene-4-sulfonate, r-+-3-aminotetrahydrofuran toluene-4-sulfonate, r---3-aminotetrahydrofurantoluene-4-sulfonate, 3-furanamine, tetrahydro-, r-, 4-methylbenzenesulfonate, c4h9no.c7h8o3s, r-tetrahydrofuran-3-amine-4-methylbenzenesulfonate, 3r-oxolan-3-amine; para-toluene sulfonate, 4-methylbenzenesulfonic acid; 3r-tetrahydrofuran-3-amine | |
| 4-methylbenzenesulfonic acid;(3R)-oxolan-3-amine |
Specifications
| 111769-27-8 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| MFCD00040622,MFCD00040622,MFCD00066238 | |
| 4.3 | |
| BZXPLADBSZWDIH-UHFFFAOYNA-N | |
| 4-methylbenzenesulfonic acid;(3R)-oxolan-3-amine | |
| 14243169 | |
| 95% |
| 96.0 | |
| 48.91 to 52.99% C, 5.18 to 5.62% N | |
| 95% | |
| C11H17NO4S | |
| 5g | |
| r-3-aminotetrahydrofuran tosylate, r-+-tetrahydro-3-furylamine p-toluenesulfonate salt, r---3-aminotetrahydrofuran toluene-4-sulfonate, r-+-3-aminotetrahydrofuran toluene-4-sulfonate, r---3-aminotetrahydrofurantoluene-4-sulfonate, 3-furanamine, tetrahydro-, r-, 4-methylbenzenesulfonate, c4h9no.c7h8o3s, r-tetrahydrofuran-3-amine-4-methylbenzenesulfonate, 3r-oxolan-3-amine; para-toluene sulfonate, 4-methylbenzenesulfonic acid; 3r-tetrahydrofuran-3-amine | |
| NC1CCOC1.CC1=CC=C(C=C1)S(O)(=O)=O | |
| 259.32 | |
| 259.32 | |
| (R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only