missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Aminoquinuclidine Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7118098
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28301G 1 g
A28305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28301G Supplier TCI America Supplier No. A28301G

Chemical Identifiers

CAS 123536-14-1
Molecular Formula C7H16Cl2N2
Molecular Weight (g/mol) 199.119
MDL Number MFCD00191752
InChI Key STZHBULOYDCZET-KLXURFKVSA-N
Synonym r-3-aminoquinuclidine dihydrochloride, r-+-3-aminoquinuclidine dihydrochloride, r-quinuclidin-3-amine dihydrochloride, 3r-1-azabicyclo 2.2.2 octan-3-amine dihydrochloride, 3r-quinuclidin-3-amine dihydrochloride, r-+-3-aminoquinuclidine 2hcl, 3r-3-aminoquinuclidine dihydrochloride, r-1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride, 3r, pubchem9967
PubChem CID 12494972
IUPAC Name (3R)-1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
SMILES C1CN2CCC1C(C2)N.Cl.Cl

Specifications

Melting Point 315°C
Color White-Yellow
Quantity 1 g
Formula Weight 199.12
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-3-Aminoquinuclidine Dihydrochloride