missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-Amino-3-phenylpropionic Acid 98.0+%, TCI America™
$55.92 - $218.10
Chemical Identifiers
| CAS | 13921-90-9 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.192 |
| MDL Number | MFCD01076237 |
| InChI Key | UJOYFRCOTPUKAK-MRVPVSSYSA-N |
| Synonym | D-beta-Phenylalanine |
| PubChem CID | 686703 |
| ChEBI | CHEBI:67172 |
| IUPAC Name | (3R)-3-amino-3-phenylpropanoic acid |
| SMILES | C1=CC=C(C=C1)C(CC(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2655200MG
|
TCI America
A2655200MG |
200 mg |
Each for $55.92
|
|
|||||
|
A26551G
|
TCI America
A26551G |
1 g |
Each for $218.10
|
|
|||||
Chemical Identifiers
| 13921-90-9 | |
| 165.192 | |
| UJOYFRCOTPUKAK-MRVPVSSYSA-N | |
| 686703 | |
| (3R)-3-amino-3-phenylpropanoic acid |
| C9H11NO2 | |
| MFCD01076237 | |
| D-beta-Phenylalanine | |
| CHEBI:67172 | |
| C1=CC=C(C=C1)C(CC(=O)O)N |
Specifications
| 211°C | |
| White | |
| MFCD01076237 | |
| UJOYFRCOTPUKAK-MRVPVSSYSA-N | |
| (3R)-3-amino-3-phenylpropanoic acid | |
| 165.192 | |
| CHEBI:67172 | |
| ≥98.0% (HPLC,T) | |
| (R)-3-Amino-3-phenylpropionic Acid |
| 13921-90-9 | |
| C9H11NO2 | |
| D-beta-Phenylalanine | |
| C1=CC=C(C=C1)C(CC(=O)O)N | |
| 200 mg | |
| 686703 | |
| 165.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No