missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, 98% ee

Catalog No. p-57642
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC362990010 1 g Glass Bottle
AC362990050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC362990010 Supplier Thermo Scientific Chemicals Supplier No. 362990010

CAS: 340188-50-3 | C11H15NO2·HCl | 229.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 340188-50-3
Molecular Formula C11H15NO2·HCl
Molecular Weight (g/mol) 229.7
MDL Number MFCD02259750
InChI Key ATSZQDTVNRNXKB-HNCPQSOCSA-N
Synonym r-3-amino-3-phenylpropanoic acid ethyl ester hydrochloride, r-ethyl 3-amino-3-phenylpropanoate hydrochloride, ethyl 3r-3-amino-3-phenylpropanoate hydrochloride, r-3-amino-3-phenylpropanoic acid ethyl ester hcl, 3r-3-amino-3-phenylpropanoic acid ethyl ester hcl, pubchem13851, r-beta-phenylalanine ethyl ester hydrochloride, r-3-amino-3-phenylpropanoic acid ethyl ester hyd, r-3-amino-3-phenylpropionic acid ethyl ester hcl, r-3-amino-3-phenyl-propionic acid ethyl ester hydro chloride
PubChem CID 2734508
IUPAC Name ethyl (3R)-3-amino-3-phenylpropanoate;hydrochloride
SMILES CCOC(=O)CC(C1=CC=CC=C1)N.Cl

Specifications

Melting Point 119°C
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (GC)
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 229.7
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, (98% e.e.)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only