missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-3-phenyl-1-propanol 97.0+%, TCI America™

Catalog No. A2999200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2999200MG 200 mg
A29991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2999200MG Supplier TCI America Supplier No. A2999200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 170564-98-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD01311791
InChI Key SEQXIQNPMQTBGN-SECBINFHSA-N
Synonym r-1-phenyl-3-propanolamine, r-3-amino-3-phenylpropan-1-ol, 3r-3-amino-3-phenylpropan-1-ol, r-beta-phenylalaninol, r-3-amino-3-phenyl-1-propanol, r-3-phenyl-beta-alaninol, r-3-amino-3-phenyl-propan-1-ol, r--phenylalaninol, pubchem13853
PubChem CID 7016858
IUPAC Name (3R)-3-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCO)N

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 200 mg
UN Number 3259
Formula Weight 151.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (R)-3-Amino-3-phenyl-1-propanol
TSCA No
Recommended Storage Refrigerator