missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-1-butanol 98.0+%, TCI America™

Catalog No. A29511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29511G 1 g
A29515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29511G Supplier TCI America Supplier No. A29511G
Only null left

Chemical Identifiers

CAS 61477-40-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD13184351
InChI Key AGMZSYQMSHMXLT-SCSAIBSYSA-N
PubChem CID 9898801
IUPAC Name (3R)-3-aminobutan-1-ol
SMILES CC(CCO)N

Specifications

Color Yellow
Quantity 1 g
UN Number 2734
Formula Weight 89.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-3-Amino-1-butanol