missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-1-benzylpiperidine, 97%

Catalog No. p-7034727
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5242503 1 g
AAH5242506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5242503 Supplier Thermo Scientific Chemicals Supplier No. H5242503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 168466-84-0 | C12H18N2 | 190.29 g/mol

(R)-3-Amino-1-benzylpiperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-Amino-1-benzylpiperidine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 168466-84-0
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD03093374
InChI Key HARWNWOLWMTQCC-GFCCVEGCSA-N
Synonym r-3-amino-1-benzylpiperidine, r-1-benzylpiperidin-3-amine, r-3-amino-1-benzyl-piperidine, 3r-1-benzylpiperidin-3-amine, r-1-benzyl-3-aminopiperidine, 3-piperidinamine, 1-phenylmethyl-, 3r, s-3-amino-n-benzylpiperidine, pubchem11493, r-n-benzyl-3-aminopiperidine, r-1-benzyl-piperidin-3-ylamine
PubChem CID 854130
IUPAC Name (3R)-1-benzylpiperidin-3-amine
SMILES C1CC(CN(C1)CC2=CC=CC=C2)N

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material (R)-3-Amino-1-benzylpiperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only