missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-Amino-1,2-propanediol 98.0+%, TCI America™
$39.14 - $109.98
Chemical Identifiers
| CAS | 66211-46-9 |
|---|---|
| Molecular Formula | C3H9NO2 |
| Molecular Weight (g/mol) | 91.11 |
| MDL Number | MFCD00798261 |
| InChI Key | KQIGMPWTAHJUMN-GSVOUGTGSA-N |
| Synonym | r-3-amino-1,2-propanediol, r-3-aminopropane-1,2-diol, r-+-3-amino-1,2-propandiol, 2r-3-aminopropane-1,2-diol, r-+-3-amino-1,2-propanediol, r-+-amino-1,2-propanediol, 1-aminoglycerol, 1,2-propanediol, 3-amino-, 2r, pubchem5711, r-3-amino-1,2-propanol |
| PubChem CID | 6994409 |
| IUPAC Name | (2R)-3-aminopropane-1,2-diol |
| SMILES | C(C(CO)O)N |
Chemical Identifiers
| 66211-46-9 | |
| 91.11 | |
| KQIGMPWTAHJUMN-GSVOUGTGSA-N | |
| 6994409 | |
| C(C(CO)O)N |
| C3H9NO2 | |
| MFCD00798261 | |
| r-3-amino-1,2-propanediol, r-3-aminopropane-1,2-diol, r-+-3-amino-1,2-propandiol, 2r-3-aminopropane-1,2-diol, r-+-3-amino-1,2-propanediol, r-+-amino-1,2-propanediol, 1-aminoglycerol, 1,2-propanediol, 3-amino-, 2r, pubchem5711, r-3-amino-1,2-propanol | |
| (2R)-3-aminopropane-1,2-diol |
Specifications
| 66211-46-9 | |
| White-Yellow | |
| C3H9NO2 | |
| 1 g | |
| r-3-amino-1,2-propanediol, r-3-aminopropane-1,2-diol, r-+-3-amino-1,2-propandiol, 2r-3-aminopropane-1,2-diol, r-+-3-amino-1,2-propanediol, r-+-amino-1,2-propanediol, 1-aminoglycerol, 1,2-propanediol, 3-amino-, 2r, pubchem5711, r-3-amino-1,2-propanol | |
| C(C(CO)O)N | |
| 91.11 | |
| 91.11 | |
| Crystalline Powder |
| 55°C | |
| 163°C | |
| MFCD00798261 | |
| 3259 | |
| KQIGMPWTAHJUMN-GSVOUGTGSA-N | |
| (2R)-3-aminopropane-1,2-diol | |
| 6994409 | |
| ≥98.0% (GC) | |
| (R)-3-Amino-1,2-propanediol |
Safety and Handling
EINECSNumber : (2)-3504
RTECSNumber : TY2800000
TSCA : No