missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine 98.0+%, TCI America™
$104.38 - $335.52
Chemical Identifiers
| CAS | 157488-65-8 |
|---|---|
| Molecular Formula | C22H18NO2P |
| Molecular Weight (g/mol) | 359.365 |
| MDL Number | MFCD03426988 |
| InChI Key | QCHAVHXSBZARBO-UHFFFAOYSA-N |
| PubChem CID | 10893715 |
| SMILES | CN(C)P1OC2=C(C3=CC=CC=C3C=C2)C4=C(O1)C=CC5=CC=CC=C54 |
Chemical Identifiers
| 157488-65-8 | |
| 359.365 | |
| QCHAVHXSBZARBO-UHFFFAOYSA-N | |
| CN(C)P1OC2=C(C3=CC=CC=C3C=C2)C4=C(O1)C=CC5=CC=CC=C54 |
| C22H18NO2P | |
| MFCD03426988 | |
| 10893715 |
Specifications
| 157488-65-8 | |
| White-Yellow | |
| MFCD03426988 | |
| QCHAVHXSBZARBO-UHFFFAOYSA-N | |
| 359.365 | |
| 359.36 | |
| Crystalline Powder |
| 227°C | |
| C22H18NO2P | |
| 1 g | |
| CN(C)P1OC2=C(C3=CC=CC=C3C=C2)C4=C(O1)C=CC5=CC=CC=C54 | |
| 10893715 | |
| ≥98.0% (HPLC,N) | |
| (R)-(-)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4-a′]dinaphthalen-4-yl)dimethylamine |
Safety and Handling
TSCA : No