missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine 98.0+%, TCI America™

Catalog No. D49925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49925G 5 g
D49921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49925G Supplier TCI America Supplier No. D49925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 157488-65-8
Molecular Formula C22H18NO2P
Molecular Weight (g/mol) 359.365
MDL Number MFCD03426988
InChI Key QCHAVHXSBZARBO-UHFFFAOYSA-N
PubChem CID 10893715
SMILES CN(C)P1OC2=C(C3=CC=CC=C3C=C2)C4=C(O1)C=CC5=CC=CC=C54

Specifications

Melting Point 227°C
Color White-Yellow
Quantity 5 g
Formula Weight 359.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4-a′]dinaphthalen-4-yl)dimethylamine