missing translation for 'onlineSavingsMsg'
Learn More

(R)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl 98.0+%, TCI America™

Catalog No. p-7127508
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2801200MG 200 mg
D28011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2801200MG Supplier TCI America Supplier No. D2801200MG

Chemical Identifiers

CAS 75714-59-9
Molecular Formula C22H16Br2O2
Molecular Weight (g/mol) 472.176
MDL Number MFCD04038415
InChI Key DFTUKDIMHCCQIT-UHFFFAOYSA-N
PubChem CID 394170
IUPAC Name 3-bromo-1-(3-bromo-2-methoxynaphthalen-1-yl)-2-methoxynaphthalene
SMILES COC1=C(C2=CC=CC=C2C=C1Br)C3=C(C(=CC4=CC=CC=C43)Br)OC

Specifications

Melting Point 185°C
Color White
Quantity 200 mg
Formula Weight 472.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-3,3′-Dibromo-2,2′-dimethoxy-1,1′-binaphthyl