missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Pyrrolidinone-5-carboxylic acid, 98+%

Catalog No. AAB2467806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2467806 5 g
AAB2467814 25 g
AAB2467822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2467806 Supplier Thermo Scientific Chemicals Supplier No. B2467806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4042-36-8 | C5H7NO3 | 129.115 g/mol

(R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing /reducing agents, strong acids, strong bases.

Chemical Identifiers

CAS 4042-36-8
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
MDL Number MFCD00066212
InChI Key ODHCTXKNWHHXJC-GSVOUGTGSA-N
Synonym d-pyroglutamic acid, 5-oxo-d-proline, r-5-oxopyrrolidine-2-carboxylic acid, d-pyr-oh, 2r-5-oxopyrrolidine-2-carboxylic acid, d-proline, 5-oxo, r-+-2-pyrrolidone-5-carboxylic acid, d-5-pyrrolidone-2-carboxylic acid, r-2-pyrrolidone-5-carboxylic acid, d-5-oxo-2-pyrrolidinecarboxylic acid
PubChem CID 439685
ChEBI CHEBI:16924
IUPAC Name (2R)-5-oxopyrrolidine-2-carboxylic acid
SMILES C1CC(=O)NC1C(=O)O

Specifications

Melting Point 158°C to 160°C
Quantity 5 g
Beilstein 82133
Solubility Information Soluble in water.
Optical Rotation +10.5° (c=5 in Water)
Formula Weight 129.12
Percent Purity ≥98%
Chemical Name or Material (R)-(+)-2-Pyrrolidinone-5-carboxylic acid
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 223-735-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only