missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Phenylpropionic Acid 98.0+%, TCI America™

Catalog No. p-7135015
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
P12191G 1 g
P12195G 5 g
2 options

Catalog No. P12191G

Supplier: TCI America P12191G

Only null left

Chemical Identifiers

CAS 7782-26-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00063140
InChI Key YPGCWEMNNLXISK-SSDOTTSWSA-N
Synonym r---2-phenylpropionic acid, r-2-phenylpropanoic acid, 2r-2-phenylpropanoic acid, r-hydratropic acid, --hydratropic acid, r---hydratropic acid, r---2-phenylpropionicacid, r-2-phenylpropionic acid, r-2-phenyl-propionic acid, r-2-phenyl-proprionic acid
PubChem CID 446626
ChEBI CHEBI:43035
IUPAC Name (2R)-2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-2-Phenylpropionic Acid
EINECSNumber (3)-1751
TSCA No