missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Phenylpropionic acid, 97%

Catalog No. AAA1763901
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1763901 0.25 g
AAA1763903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1763901 Supplier Thermo Scientific Chemicals Supplier No. A1763901
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7782-26-5 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-26-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00063140
InChI Key YPGCWEMNNLXISK-SSDOTTSWSA-N
Synonym r---2-phenylpropionic acid, r-2-phenylpropanoic acid, 2r-2-phenylpropanoic acid, r-hydratropic acid, --hydratropic acid, r---hydratropic acid, r---2-phenylpropionicacid, r-2-phenylpropionic acid, r-2-phenyl-propionic acid, r-2-phenyl-proprionic acid
PubChem CID 446626
ChEBI CHEBI:43035
IUPAC Name (2R)-2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 32°C to 34°C
Density 1.1
Boiling Point 264°C to 266°C
Flash Point 148°C (298°F)
Refractive Index 1.523
Quantity 0.25 g
Beilstein 2207688
Optical Rotation −73° (c=1.6 in Chloroform)
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material (R)-(-)-2-Phenylpropionic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only