missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Pentanol 98.0+%, TCI America™

Catalog No. p-7134745
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07441ML 1 mL
P07445ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07441ML Supplier TCI America Supplier No. P07441ML
Only null left

Chemical Identifiers

CAS 31087-44-2
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00065953
InChI Key JYVLIDXNZAXMDK-RXMQYKEDSA-N
Synonym r---2-pentanol, 2r-pentan-2-ol, r-2-pentanol, 2r-2-pentanol, 2-pentanol #, r-pentan-2-ol, pubchem6721, 2-pentanol, 2r
PubChem CID 7014876
IUPAC Name (2R)-pentan-2-ol
SMILES CCCC(C)O

Specifications

Color Colorless
Boiling Point 119°C
Quantity 1 mL
UN Number 1105
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-2-Pentanol
EINECSNumber (2)-0217
RTECSNumber SA4900000
TSCA No