missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2-Octanol 98.0+%, TCI America™
$231.64 - $689.20
Chemical Identifiers
| CAS | 5978-70-1 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.231 |
| MDL Number | MFCD00064284 |
| InChI Key | SJWFXCIHNDVPSH-MRVPVSSYSA-N |
| Synonym | r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol |
| PubChem CID | 80080 |
| ChEBI | CHEBI:37871 |
| IUPAC Name | (2R)-octan-2-ol |
| SMILES | CCCCCCC(C)O |
Chemical Identifiers
| 5978-70-1 | |
| 130.231 | |
| SJWFXCIHNDVPSH-MRVPVSSYSA-N | |
| 80080 | |
| (2R)-octan-2-ol |
| C8H18O | |
| MFCD00064284 | |
| r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol | |
| CHEBI:37871 | |
| CCCCCCC(C)O |
Specifications
| 5978-70-1 | |
| C8H18O | |
| 5 mL | |
| SJWFXCIHNDVPSH-MRVPVSSYSA-N | |
| (2R)-octan-2-ol | |
| 80080 | |
| 130.23 | |
| Liquid |
| Colorless | |
| MFCD00064284 | |
| r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol | |
| CCCCCCC(C)O | |
| 130.231 | |
| CHEBI:37871 | |
| ≥98.0% (GC) | |
| (R)-(-)-2-Octanol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes