missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Octanol 98.0+%, TCI America™

Catalog No. O01455ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01455ML 5 mL
O014525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01455ML Supplier TCI America Supplier No. O01455ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5978-70-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00064284
InChI Key SJWFXCIHNDVPSH-MRVPVSSYSA-N
Synonym r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
PubChem CID 80080
ChEBI CHEBI:37871
IUPAC Name (2R)-octan-2-ol
SMILES CCCCCCC(C)O

Specifications

Color Colorless
Quantity 5 mL
Formula Weight 130.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-2-Octanol
EINECSNumber (2)-0217
TSCA Yes