missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Octanol, 99%

Catalog No. AAL1150206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1150206 5 g
AAL1150203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1150206 Supplier Thermo Scientific Chemicals Supplier No. L1150206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5978-70-1 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5978-70-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00064284
InChI Key SJWFXCIHNDVPSH-MRVPVSSYSA-N
Synonym r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
PubChem CID 80080
ChEBI CHEBI:37871
IUPAC Name (2R)-octan-2-ol
SMILES CCCCCCC(C)O

Specifications

Density 0.819
Boiling Point 179°C to 180°C
Flash Point 76°C (169°F)
Odor Pleasant
Refractive Index 1.426
Quantity 5 g
Beilstein 1719324
Merck Index 14,6752
Optical Rotation −10° (c=1 in Ethanol)
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material (R)-(-)-2-Octanol
Show More Show Less
Hazard Category H226-H301-H315-H319-H335
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 227-777-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only