missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2-Methylpiperazine 98.0+%, TCI America™
$229.10 - $727.72
Chemical Identifiers
| CAS | 75336-86-6 |
|---|---|
| Molecular Formula | C5H12N2 |
| Molecular Weight (g/mol) | 100.165 |
| MDL Number | MFCD00192317 |
| InChI Key | JOMNTHCQHJPVAZ-RXMQYKEDSA-N |
| Synonym | r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w |
| PubChem CID | 7330434 |
| IUPAC Name | (2R)-2-methylpiperazine |
| SMILES | CC1CNCCN1 |
Chemical Identifiers
| 75336-86-6 | |
| 100.165 | |
| JOMNTHCQHJPVAZ-RXMQYKEDSA-N | |
| 7330434 | |
| CC1CNCCN1 |
| C5H12N2 | |
| MFCD00192317 | |
| r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w | |
| (2R)-2-methylpiperazine |
Specifications
| 75336-86-6 | |
| White-Yellow | |
| C5H12N2 | |
| 5 g | |
| r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w | |
| CC1CNCCN1 | |
| 100.165 | |
| 100.17 | |
| Crystalline Powder |
| 90°C | |
| 155°C | |
| MFCD00192317 | |
| 2925 | |
| JOMNTHCQHJPVAZ-RXMQYKEDSA-N | |
| (2R)-2-methylpiperazine | |
| 7330434 | |
| ≥98.0% (GC,T) | |
| (R)-(-)-2-Methylpiperazine |
Safety and Handling
EINECSNumber : (5)-3829
RTECSNumber : TM1225020
TSCA : No