missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Methylpiperazine, 95%

Catalog No. p-4510661
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC356270050 5 g
AC356270250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC356270050 Supplier Thermo Scientific Chemicals Supplier No. 356270050
Only null left

CAS: 75336-86-6 | C5H12N2 | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75336-86-6
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.16
InChI Key JOMNTHCQHJPVAZ-RXMQYKEDSA-N
Synonym r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w
PubChem CID 7330434
IUPAC Name (2R)-2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Melting Point 86.0°C to 91.0°C
Color White to Yellow
Boiling Point 155.0°C to 156.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition − 18.00 (20.00°C c=1,toluene)
Specific Rotation − 18.00
Solubility Information Solubility in water: soluble.
Formula Weight 100.16
Percent Purity 95%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material (R)-(-)-2-Methylpiperazine
Show More Show Less
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only