missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Methylpiperazine, 98+%

Catalog No. AAB2236606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2236606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2236606 Supplier Thermo Scientific Chemicals Supplier No. B2236606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75336-86-6 | C5H12N2 | 100.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75336-86-6
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00192317
InChI Key JOMNTHCQHJPVAZ-RXMQYKEDSA-N
Synonym r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w
PubChem CID 7330434
IUPAC Name (2R)-2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Melting Point 76°C to 78°C
Boiling Point 155°C to 156°C
Flash Point 65°C (149°F)
Quantity 5 g
UN Number UN2925
Beilstein 3647994
Sensitivity Air sensitive; Hygroscopic
Optical Rotation −7° (c=1 in Ethanol)
Formula Weight 100.17
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-2-Methylpiperazine
Show More Show Less
Hazard Category H228-H314
Hazard Statement GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only