missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Methoxymethyl)pyrrolidine 99.0+%, TCI America™

Catalog No. p-7132573
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11691G 1 g
M11695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11691G Supplier TCI America Supplier No. M11691G
Only null left

Chemical Identifiers

CAS 84025-81-0
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00066219
InChI Key CHPRFKYDQRKRRK-ZCFIWIBFSA-N
Synonym r-2-methoxymethyl pyrrolidine, 2r-2-methoxymethyl pyrrolidine, r---2-methoxymethyl pyrrolidine, o-methyl-d-prolinol, r-2-methoxymethyl-pyrrolidine, r-2-methoxymethylpyrrolidine, pyrrolidine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl-pyrrolidine, r-+-2-methoxymethyl pyrrolidine, h-pro-ol-me
PubChem CID 671216
IUPAC Name (2R)-2-(methoxymethyl)pyrrolidine
SMILES COCC1CCCN1

Specifications

Color Colorless
Quantity 1 g
UN Number 1993
Formula Weight 115.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-2-(Methoxymethyl)pyrrolidine