missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98+%

Catalog No. AAL19492MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19492MD 250 mg
AAL1949203 1 g
AAL1949206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL19492MD Supplier Thermo Scientific Chemicals Supplier No. L19492MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84025-81-0 | C6H13NO | 115.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84025-81-0
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00066219
InChI Key CHPRFKYDQRKRRK-ZCFIWIBFSA-N
Synonym r-2-methoxymethyl pyrrolidine, 2r-2-methoxymethyl pyrrolidine, r---2-methoxymethyl pyrrolidine, o-methyl-d-prolinol, r-2-methoxymethyl-pyrrolidine, r-2-methoxymethylpyrrolidine, pyrrolidine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl-pyrrolidine, r-+-2-methoxymethyl pyrrolidine, h-pro-ol-me
PubChem CID 671216
IUPAC Name (2R)-2-(methoxymethyl)pyrrolidine
SMILES COCC1CCCN1

Specifications

Density 0.933
Boiling Point 61°C to 62°C (40 mmHg)
Flash Point 45°C (113°F)
Refractive Index 1.4457
Quantity 250 mg
UN Number UN1993
Beilstein 4229755
Sensitivity Air Sensitive
Optical Rotation −2.2° (c=2 in Benzene)
Formula Weight 115.18
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-2-(Methoxymethyl)pyrrolidine
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only