missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2-Isopropylamino-2-phenylethanol 98.0+%, TCI America™
$402.36 - $1,209.00
Chemical Identifiers
| CAS | 112211-92-4 |
|---|---|
| Molecular Formula | C11H17NO |
| Molecular Weight (g/mol) | 179.26 |
| MDL Number | MFCD00671548 |
| InChI Key | XFSCVCVPECOHBL-UHFFFAOYNA-N |
| PubChem CID | 12664771 |
| IUPAC Name | 2-phenyl-2-[(propan-2-yl)amino]ethan-1-ol |
| SMILES | CC(C)NC(CO)C1=CC=CC=C1 |
Chemical Identifiers
| 112211-92-4 | |
| 179.26 | |
| XFSCVCVPECOHBL-UHFFFAOYNA-N | |
| 2-phenyl-2-[(propan-2-yl)amino]ethan-1-ol |
| C11H17NO | |
| MFCD00671548 | |
| 12664771 | |
| CC(C)NC(CO)C1=CC=CC=C1 |
Specifications
| 112211-92-4 | |
| White-Yellow | |
| MFCD00671548 | |
| XFSCVCVPECOHBL-UHFFFAOYNA-N | |
| 2-phenyl-2-[(propan-2-yl)amino]ethan-1-ol | |
| 12664771 | |
| ≥98.0% (GC,T) | |
| (R)-2-Isopropylamino-2-phenylethanol |
| 77°C | |
| C11H17NO | |
| 5 g | |
| CC(C)NC(CO)C1=CC=CC=C1 | |
| 179.26 | |
| 179.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes