Learn More
(R)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% ee)
CAS: 130165-76-3 | C9H16O4 | 188.22 g/mol
$239.02 - $321.28
Chemical Identifiers
| CAS | 130165-76-3 |
|---|---|
| Molecular Formula | C9H16O4 |
| Molecular Weight (g/mol) | 188.22 |
| MDL Number | MFCD01091032 |
| InChI Key | AJNNFMRCMKGYHA-SSDOTTSWSA-N |
| Synonym | r-2-isobutylsuccinic acid-1-methyl ester, 3r-3-methoxycarbonyl-5-methylhexanoic acid, r-3-methoxycarbonyl-5-methylhexanoic acid, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2r, r-2-isobutyl succinic acid 1-methyl ester, r-+-2-isobutylsuccinic acid 1-methyl ester |
| PubChem CID | 5702633 |
| IUPAC Name | (3R)-3-methoxycarbonyl-5-methylhexanoic acid |
| SMILES | CC(C)CC(CC(=O)O)C(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC363055000
|
Thermo Scientific Chemicals
363055000 |
500 mg | Glass Bottle |
Each for $239.02
|
|
||||
|
AC363050010
|
Thermo Scientific Chemicals
363050010 |
1 g | Glass bottle |
Each for $321.28
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 130165-76-3 | |
| 188.22 | |
| AJNNFMRCMKGYHA-SSDOTTSWSA-N | |
| 5702633 | |
| CC(C)CC(CC(=O)O)C(=O)OC |
| C9H16O4 | |
| MFCD01091032 | |
| r-2-isobutylsuccinic acid-1-methyl ester, 3r-3-methoxycarbonyl-5-methylhexanoic acid, r-3-methoxycarbonyl-5-methylhexanoic acid, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2r, r-2-isobutyl succinic acid 1-methyl ester, r-+-2-isobutylsuccinic acid 1-methyl ester | |
| (3R)-3-methoxycarbonyl-5-methylhexanoic acid |
Specifications
| 130165-76-3 | |
| 97.5% min. (GC) | |
| Glass Bottle | |
| MFCD01091032 | |
| +18° to +21° (25°C, 589nm) (c=1, CH3OH) | |
| AJNNFMRCMKGYHA-SSDOTTSWSA-N | |
| (3R)-3-methoxycarbonyl-5-methylhexanoic acid | |
| 5702633 | |
| 95%,(98% ee) | |
| (R)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% e.e.) |
| Yellow | |
| Authentic | |
| C9H16O4 | |
| 500 mg | |
| r-2-isobutylsuccinic acid-1-methyl ester, 3r-3-methoxycarbonyl-5-methylhexanoic acid, r-3-methoxycarbonyl-5-methylhexanoic acid, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2r, r-2-isobutyl succinic acid 1-methyl ester, r-+-2-isobutylsuccinic acid 1-methyl ester | |
| CC(C)CC(CC(=O)O)C(=O)OC | |
| 188.22 | |
| 188.22 | |
| Viscous Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove
GHS Signal Word: Warning
TSCA : TSCA
RUO – Research Use Only