missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Chloromandelic acid, ChiPros 99+%, ee 99+%

Catalog No. p-7047662
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1958603 1 g
AAL1958606 5 g
AAL1958614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1958603 Supplier Thermo Scientific Chemicals Supplier No. L1958603
Only null left

CAS: 52950-18-2 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52950-18-2
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00798429
InChI Key RWOLDZZTBNYTMS-SSDOTTSWSA-N
Synonym r---2-chloromandelic acid, r-2-2-chlorophenyl-2-hydroxyacetic acid, r-2-chloromandelic acid, r-o-chloromandelic acid, 2-chloro-d-mandelic acid, 2r-2-2-chlorophenyl-2-hydroxyacetic acid, r-2-chlorophenyl hydroxy acetic acid, pubchem5742, ksc914s2l, r---2-chloromandelicacid
PubChem CID 2733641
IUPAC Name (2R)-2-(2-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC=C(C(=C1)C(C(=O)O)O)Cl

Specifications

Melting Point 119°C to 121°C
Density 1.48
Quantity 1 g
Beilstein 2693370
Formula Weight 186.6
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (R)-(-)-2-Chloromandelic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H318-H317
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 444-040-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only