missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Chlorobutyric Acid 98.0+%, TCI America™

Catalog No. C21091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21091G 1 g
C21095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21091G Supplier TCI America Supplier No. C21091G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54053-45-1
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00067098
InChI Key RVBUZBPJAGZHSQ-GSVOUGTGSA-N
PubChem CID 5326288
IUPAC Name (2R)-2-chlorobutanoic acid
SMILES CCC(C(=O)O)Cl

Specifications

Color Yellow
Boiling Point 98°C
Quantity 1 g
UN Number 3265
Formula Weight 122.55
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (R)-2-Chlorobutyric Acid