missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Bromopropionic Acid 98.0+%, TCI America™

Catalog No. b17565g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B17565G 5 g
B175625G 25 g
2 options

Catalog No. B17565G

Supplier: TCI America B17565G

Only null left

Chemical Identifiers

CAS 10009-70-8
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.975
MDL Number MFCD00145205
InChI Key MONMFXREYOKQTI-UWTATZPHSA-N
PubChem CID 6992152
IUPAC Name (2R)-2-bromopropanoic acid
SMILES CC(C(=O)O)Br

Specifications

Boiling Point 104°C
Quantity 5 g
UN Number 3265
Synonym r-+-2-bromopropionic acid, r-2-bromopropanoic acid, 2r-2-bromopropanoic acid, propanoic acid,2-bromo-, 2r, propanoic acid, 2-bromo-, 2r, r-bromopropionic acid, r-2-bromopropionic acid, r-2-bromo-propionic acid, 2r-2-bromo-propionic acid, r-+-2-bromopropionicacid
Formula Weight 152.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-2-Bromopropionic Acid
EINECSNumber (9)-1167
TSCA No