missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate, Thermo Scientific™

Chemical Identifiers

CAS 1301706-76-2
Molecular Formula C19H34N2O8
Molecular Weight (g/mol) 418.487
MDL Number MFCD09953465
InChI Key HCNKZGJFFYKTGU-SNXORLAUSA-N
Synonym r-+-2-boc-aminomethyl-4,4-diethoxypiperidine fumarate, 2r-2-aminomethyl-4,4-diethoxypiperidine fumarate, 2-boc protected, fumaric acid; tert-butyl n-2r-4,4-diethoxypiperidin-2-yl methyl carbamate, tert-butyl 2r-4,4-diethoxypiperidin-2-ylmethyl carbamate 2e-but-2-enedioate
PubChem CID 44119499
IUPAC Name (E)-but-2-enedioic acid;tert-butyl N-[[(2R)-4,4-diethoxypiperidin-2-yl]methyl]carbamate
SMILES CCOC1(CCNC(C1)CNC(=O)OC(C)(C)C)OCC.C(=CC(=O)O)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH25909ME
View Documents Promotion Details
N/A 500mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1301706-76-2
418.487
HCNKZGJFFYKTGU-SNXORLAUSA-N
44119499
CCOC1(CCNC(C1)CNC(=O)OC(C)(C)C)OCC.C(=CC(=O)O)C(=O)O
C19H34N2O8
MFCD09953465
r-+-2-boc-aminomethyl-4,4-diethoxypiperidine fumarate, 2r-2-aminomethyl-4,4-diethoxypiperidine fumarate, 2-boc protected, fumaric acid; tert-butyl n-2r-4,4-diethoxypiperidin-2-yl methyl carbamate, tert-butyl 2r-4,4-diethoxypiperidin-2-ylmethyl carbamate 2e-but-2-enedioate
(E)-but-2-enedioic acid;tert-butyl N-[[(2R)-4,4-diethoxypiperidin-2-yl]methyl]carbamate

Specifications

1301706-76-2
MFCD09953465
HCNKZGJFFYKTGU-SNXORLAUSA-N
(E)-but-2-enedioic acid;tert-butyl N-[[(2R)-4,4-diethoxypiperidin-2-yl]methyl]carbamate
44119499
(R)-(+)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate
C19H34N2O8
r-+-2-boc-aminomethyl-4,4-diethoxypiperidine fumarate, 2r-2-aminomethyl-4,4-diethoxypiperidine fumarate, 2-boc protected, fumaric acid; tert-butyl n-2r-4,4-diethoxypiperidin-2-yl methyl carbamate, tert-butyl 2r-4,4-diethoxypiperidin-2-ylmethyl carbamate 2e-but-2-enedioate
CCOC1(CCNC(C1)CNC(=O)OC(C)(C)C)OCC.C(=CC(=O)O)C(=O)O
418.487
418.49
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents