missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine 98.0+%, TCI America™

Catalog No. A27121G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27121G Supplier TCI America Supplier No. A27121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 259537-92-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03419256
InChI Key SOGXYCNKQQJEED-MRVPVSSYSA-N
Synonym r-2-aminomethyl-1-n-boc-pyrrolidine, r-1-boc-2-aminomethyl pyrrolidine, r-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2r-2-aminomethyl pyrrolidine-1-carboxylate, r-2-aminomethyl-1-boc-pyrrolidine, r-2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-2-aminomethyl-1-tert-butoxycarbonyl pyrrolidine, tert-butyl r-2-aminomethyl-1-pyrrolidinecarboxylate, r-2-aminomethyl-1-n-boc-pyyrolidine
PubChem CID 1512534
IUPAC Name tert-butyl (2R)-2-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CN

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
UN Number 2735
Formula Weight 200.28
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (R)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine
TSCA No
Recommended Storage Refrigerator