missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Aminomethyl)-1-ethylpyrrolidine, 97%

Catalog No. p-56483
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC357000050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC357000050 Supplier Thermo Scientific Chemicals Supplier No. 357000050

CAS: 22795-97-7 | C7H16N2 | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22795-97-7
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00671482
InChI Key UNRBEYYLYRXYCG-SSDOTTSWSA-N
Synonym r-1-ethylpyrrolidin-2-yl methanamine, r-+-2-aminomethyl-1-ethylpyrrolidine, r-2-aminomethyl-1-ethylpyrrolidine, 2r-1-ethylpyrrolidin-2-yl methanamine, 2-pyrrolidinemethanamine, 1-ethyl-, 2r, 2r-1-ethyl-2-pyrrolidinemethanamine, 1-ethyl-2-pyrrolidinyl methanamine #, pubchem11067, r-+-2-aminomethyl-n-ethylpyrrolidine
PubChem CID 854138
IUPAC Name [(2R)-1-ethylpyrrolidin-2-yl]methanamine
SMILES CCN1CCCC1CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9290g/mL
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4655 to 1.4675
Quantity 5 g
Specific Gravity 0.929
Specific Rotation + 99.50
Formula Weight 128.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (R)-2-(Aminomethyl)-1-ethylpyrrolidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
TSCA TSCA

RUO – Research Use Only