missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine, 95%

Catalog No. p-7038629
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
AAH57638MD 250 mg
AAH5763803 1 g
2 options

Catalog No. AAH57638MD

Supplier: Thermo Scientific Chemicals H57638MD

Only null left

CAS: 22795-97-7 | C7H16N2 | 128.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22795-97-7
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00671482
InChI Key UNRBEYYLYRXYCG-SSDOTTSWSA-N
Synonym r-1-ethylpyrrolidin-2-yl methanamine, r-+-2-aminomethyl-1-ethylpyrrolidine, r-2-aminomethyl-1-ethylpyrrolidine, 2r-1-ethylpyrrolidin-2-yl methanamine, 2-pyrrolidinemethanamine, 1-ethyl-, 2r, 2r-1-ethyl-2-pyrrolidinemethanamine, 1-ethyl-2-pyrrolidinyl methanamine #, pubchem11067, r-+-2-aminomethyl-n-ethylpyrrolidine
PubChem CID 854138
IUPAC Name [(2R)-1-ethylpyrrolidin-2-yl]methanamine
SMILES CCN1CCCC1CN

Specifications

Density 0.929
Boiling Point 50°C to 54°C
Flash Point 57°C (135°F)
Refractive Index 1.467
Quantity 250 mg
UN Number UN1993
Sensitivity Hygroscopic
Optical Rotation +95° (c=1 in Methanol)
Formula Weight 128.22
Percent Purity 95%
Chemical Name or Material (R)-(+)-2-Aminomethyl-1-ethylpyrrolidine
Show More Show Less
Hazard Category H226-H302+H312-H314-H335
Hazard Statement GHS H Statement
H301-H318-H226-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only