missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Aminomethyl-1-Boc-pyrrolidine, 97%

CAS: 259537-92-3 | C10H20N2O2 | 200.282 g/mol

Supplier:  Thermo Scientific Chemicals H5241903

 View more versions of this product

Catalog No. AAH5241903


Only null left
Explore available promotions

Description

Description

Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

(R)-2-Aminomethyl-1-Boc-pyrrolidine
98°C to 110°C (1 mmHg)
MFCD03419256
UN2735
Slightly soluble in water.
+47° (c=1 in Chloroform)
tert-butyl (2R)-2-(aminomethyl)pyrrolidine-1-carboxylate
1512534
97%
259537-92-3
C10H20N2O2
1 g
r-2-aminomethyl-1-n-boc-pyrrolidine, r-1-boc-2-aminomethyl pyrrolidine, r-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2r-2-aminomethyl pyrrolidine-1-carboxylate, r-2-aminomethyl-1-boc-pyrrolidine, r-2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-2-aminomethyl-1-tert-butoxycarbonyl pyrrolidine, tert-butyl r-2-aminomethyl-1-pyrrolidinecarboxylate, r-2-aminomethyl-1-n-boc-pyyrolidine
SOGXYCNKQQJEED-MRVPVSSYSA-N
CC(C)(C)OC(=O)N1CCCC1CN
200.282
200.28

Chemical Identifiers

259537-92-3
200.282
SOGXYCNKQQJEED-MRVPVSSYSA-N
1512534
CC(C)(C)OC(=O)N1CCCC1CN
C10H20N2O2
MFCD03419256
r-2-aminomethyl-1-n-boc-pyrrolidine, r-1-boc-2-aminomethyl pyrrolidine, r-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2r-2-aminomethyl pyrrolidine-1-carboxylate, r-2-aminomethyl-1-boc-pyrrolidine, r-2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-2-aminomethyl-1-tert-butoxycarbonyl pyrrolidine, tert-butyl r-2-aminomethyl-1-pyrrolidinecarboxylate, r-2-aminomethyl-1-n-boc-pyyrolidine
tert-butyl (2R)-2-(aminomethyl)pyrrolidine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only