missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Aminohexane, ChiPros 99+%, ee 96+%

Catalog No. AAL1930406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1930406 5 g
AAL1930414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1930406 Supplier Thermo Scientific Chemicals Supplier No. L1930406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70095-40-8 | C6H15N | 101.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70095-40-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00671627
InChI Key WGBBUURBHXLGFM-ZCFIWIBFSA-N
Synonym r-2-aminohexane, 2r-hexan-2-amine, r-2-hexylamine, 2-hexanamine, r, r-2-hexaneamine, pubchem6753, 2r-2-hexanamine, r-hexan-2-amine, 2-hexanamine, 2r, r-+-2-aminohexane
PubChem CID 6993464
IUPAC Name (2R)-hexan-2-amine
SMILES CCCCC(C)N

Specifications

Melting Point <-20°C
Density 0.755
Boiling Point 116°C
Flash Point −3°C (26°F)
Quantity 5 g
UN Number UN2733
Beilstein 1718994
Sensitivity Air Sensitive
Formula Weight 101.19
Percent Purity ≥99%
Assay ee ≥96%
Chemical Name or Material (R)-(-)-2-Aminohexane
Show More Show Less
Hazard Category H225-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 622-790-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only