missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Aminobutyric Acid 98.0+%, TCI America™

Catalog No. A13771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13771G 1 g
A13775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13771G Supplier TCI America Supplier No. A13771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2623-91-8
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00064414
InChI Key QWCKQJZIFLGMSD-GSVOUGTGSA-N
Synonym d-2-aminobutyric acid, r-2-aminobutanoic acid, d---2-aminobutyric acid, 2r-2-aminobutanoic acid, d-alpha-aminobutyric acid, r---2-aminobutyric acid, h-d-abu-oh, d--2-aminobutyric acid, d-2-aminobutyrate, d-2-aminobutanoic acid
PubChem CID 439691
ChEBI CHEBI:28797
IUPAC Name (2R)-2-aminobutanoic acid
SMILES CC[C@@H](N)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 103.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-2-Aminobutyric Acid
EINECSNumber (2)-2482
TSCA No