missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-5-phosphonopentanoic acid, 99%, Thermo Scientific Chemicals

Catalog No. AAJ64887MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64887MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64887MA Supplier Thermo Scientific Chemicals Supplier No. J64887MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A competitive NMDA antagonist

A competitive NMDA antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79055-68-8
Molecular Formula C5H12NO5P
Molecular Weight (g/mol) 197.127
MDL Number MFCD00078839
InChI Key VOROEQBFPPIACJ-SCSAIBSYSA-N
Synonym d-ap5, 5-phosphono-d-norvaline, d-norvaline, 5-phosphono, d-apv, 2r-2-amino-5-phosphonopentanoic acid, d-2-amino-5-phosphopentanoic acid, d---2-amino-5-phosphonopentanoic acid, d--apv, d-2-amino-5-phosphonovaleric acid, d--ap-5
PubChem CID 135342
IUPAC Name (2R)-2-amino-5-phosphonopentanoic acid
SMILES C(CC(C(=O)O)N)CP(=O)(O)O

Specifications

Color White
Quantity 10 mg
Sensitivity Light sensitive
Optical Rotation −22° (c=1 in 6N HCl)
Formula Weight 197.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (R)-(-)-2-Amino-5-phosphonopentanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber RD2513000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only