missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-1-propanol 98.0+%, TCI America™

Catalog No. A20025ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20025ML 5 mL
A200225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20025ML Supplier TCI America Supplier No. A20025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35320-23-1
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.111
MDL Number MFCD00064413
InChI Key BKMMTJMQCTUHRP-GSVOUGTGSA-N
Synonym d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol
PubChem CID 80308
IUPAC Name (2R)-2-aminopropan-1-ol
SMILES CC(CO)N

Specifications

Color Yellow
Boiling Point 173°C
Quantity 5 mL
UN Number 2735
Formula Weight 75.11
Percent Purity ≥98.0% (GC,T)
Grade Reagent
Physical Form Liquid
Chemical Name or Material (R)-(-)-2-Amino-1-propanol
EINECSNumber (9)-0122
TSCA No
Recommended Storage Refrigerator