missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-1-propanol, 98%, Thermo Scientific Chemicals

$336.43 - $336.43

Chemical Identifiers

CAS 35320-23-1
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064413
InChI Key BKMMTJMQCTUHRP-GSVOUGTGSA-N
Synonym d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol
PubChem CID 80308
IUPAC Name (2R)-2-aminopropan-1-ol
SMILES CC(CO)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC400440050
SDS View Documents
Thermo Scientific Chemicals
400440050
5 g Glass bottle
Each for $336.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35320-23-1
75.11
BKMMTJMQCTUHRP-GSVOUGTGSA-N
80308
CC(CO)N
C3H9NO
MFCD00064413
d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol
(2R)-2-aminopropan-1-ol

Specifications

35320-23-1
100.0
0.9650g/mL
83°C
Authentic
Glass bottle
1.4480 to 1.451
MFCD00064413
04, III, 735
− 17.00 (20.00°C c=neat)
d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol
BKMMTJMQCTUHRP-GSVOUGTGSA-N
(2R)-2-aminopropan-1-ol
80308
98%
(R)-(-)-2-Amino-1-propanol
97.5
Colorless to Yellow
173°C to 176°C
99% min. (GC)
97.5% min. (GC)
C3H9NO
CH3CH(NH2)CH2OH
5 g
0.965
-17
Solubility in water: soluble in water
CC(CO)N
75.11
75.11
Oily Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only