missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-1-propanol, 98%

Catalog No. AAL1103003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1103003 1 g
AAL1103006 5 g
AAL1103014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1103003 Supplier Thermo Scientific Chemicals Supplier No. L1103003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 35320-23-1 | C3H9NO | 75.111 g/mol

(R)-(-)-2-Amino-1-propanol is used as a chiral intermediate for the synthesis of pharmaceutical products and agrochemicals. It acts as additives in health care products and animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-2-Amino-1-propanol is used as a chiral intermediate for the synthesis of pharmaceutical products and agrochemicals. It acts as additives in health care products and animal feed.

Solubility
Miscible with water.

Notes
Air sensitive and hygroscopic. Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 35320-23-1
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.111
MDL Number MFCD00064413
InChI Key BKMMTJMQCTUHRP-GSVOUGTGSA-N
Synonym d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol
PubChem CID 80308
IUPAC Name (2R)-2-aminopropan-1-ol
SMILES CC(CO)N

Specifications

Density 0.965
Boiling Point 174°C to 176°C
Flash Point 83°C (181°F)
Refractive Index 1.4493
Quantity 1 g
UN Number UN2735
Beilstein 1718866
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Optical Rotation −22° (c=2 in Ethanol)
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (R)-(-)-2-Amino-1-propanol
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only